ENA161208640 Enamine

3-[(2-methylpropyl)amino]-1lambda6-thiolane-1,1-dione hydrochloride

  • MDL number MFCD00456645
  • Cas Number : 464876-48-0
  • Molecular Weight : 227.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-07925

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07925
Availability details for
Quantity
  • Availability
  • Add to Cart