ENA161208641 Enamine

3-(propylamino)-1lambda6-thiolane-1,1-dione hydrochloride

  • MDL number MFCD07286701
  • Cas Number : 483967-52-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118-119
Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07926

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07926
Availability details for
Quantity
  • Availability
  • Add to Cart