ENA161208643 Enamine

N-(3-fluorophenyl)-2-(methylamino)acetamide hydrochloride

  • MDL number MFCD07286700
  • Cas Number : 1049774-86-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-07940

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07940
Availability details for
Quantity
  • Availability
  • Add to Cart