ENA161208653 Enamine

4-hydrazinobenzenesulfonamide hydrochloride

  • MDL number MFCD00052262
  • Cas Number : 17852-52-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 195...197
Purity 95.00
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08026

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-08026
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart