ENA161208662 Enamine

1-(1-adamantyl)ethanamine hydrochloride

  • MDL number MFCD00072023
  • Cas Number : 1501-84-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 378...380
Purity 95.00
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08051

Safety Information

safety_statement 26-36
msds https://www.enaminestore.com/catalog/EN300-08051
Availability details for
Quantity
  • Availability
  • Add to Cart