ENA161208667 Enamine

1-(4-nitrobenzenesulfonyl)piperazine hydrochloride

  • MDL number MFCD07286044
  • Cas Number : 1049751-39-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100-101
Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08270

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08270
Availability details for
Quantity
  • Availability
  • Add to Cart