ENA161208685 Enamine

4-[1-(4-fluorophenyl)-2,5-dimethyl-1H-pyrrol-3-yl]-1,3-thiazol-2-amine hydrochloride

  • MDL number MFCD08236877
  • Cas Number : 1156039-78-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08650

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08650
Availability details for
Quantity
  • Availability
  • Add to Cart