ENA161208689 Enamine

5-chloro-2-(piperidin-1-yl)aniline hydrochloride

  • MDL number MFCD03164643
  • Cas Number : 856846-59-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08690

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08690
Availability details for
Quantity
  • Availability
  • Add to Cart