ENA161208692 Enamine

1-(2,4-dimethylbenzenesulfonyl)piperazine hydrochloride

  • MDL number MFCD03028860
  • Cas Number : 1172964-57-6
  • Molecular Weight : 290.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180-181
Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-04671

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04671
Availability details for
Quantity
  • Availability
  • Add to Cart