ENA161208694 Enamine

3-(3,5-dimethyl-1H-pyrazol-1-yl)-4H-1,2,4-triazol-4-amine hydrochloride

  • MDL number MFCD01347509
  • Cas Number : 121378-82-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125-126
Purity 95
Salt Data HCl
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08735

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08735
Availability details for
Quantity
  • Availability
  • Add to Cart