ENA161208697 Enamine

3-(phenylsulfanyl)propan-1-amine hydrochloride

  • MDL number MFCD08447166
  • Cas Number : 78540-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140-141
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-08763

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08763
Availability details for
Quantity
  • Availability
  • Add to Cart