ENA161208698 Enamine

2-(methylamino)-N-[4-(morpholin-4-yl)phenyl]acetamide dihydrochloride

  • MDL number MFCD07286200
  • Cas Number : 1173250-08-2
  • Molecular Weight : 322.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-08785

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08785
Availability details for
Quantity
  • Availability
  • Add to Cart