ENA161208712 Enamine

5-chloro-2-(4-chloro-2,6-dimethylphenoxy)aniline hydrochloride

  • MDL number MFCD08447204
  • Cas Number : 1049784-98-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190-191
Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08905

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08905
Availability details for
Quantity
  • Availability
  • Add to Cart