ENA161208717 Enamine

2-(tert-butylamino)-N-(4-fluorophenyl)acetamide hydrochloride

  • MDL number MFCD07287067
  • Cas Number : 1049751-65-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09054

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09054
Availability details for
Quantity
  • Availability
  • Add to Cart