ENA161208725 Enamine

N-(2,3-dimethylphenyl)-2-({[(2,3-dimethylphenyl)carbamoyl]methyl}amino)acetamide hydrochloride

  • MDL number MFCD07287164
  • Cas Number : 1049756-66-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09168

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09168
Availability details for
Quantity
  • Availability
  • Add to Cart