ENA161208726 Enamine

2-[(propan-2-yl)amino]acetamide hydrochloride

  • MDL number MFCD07287166
  • Cas Number : 429671-05-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09187

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09187
Availability details for
Quantity
  • Availability
  • Add to Cart