ENA161208727 Enamine

N-(2,3-dimethylphenyl)-2-[(tetrahydrofuran-2-ylmethyl)amino]acetamide hydrochloride

  • MDL number MFCD07287175
  • Cas Number : 1049768-94-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 204...206
Purity 95.00
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09191

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09191
Availability details for
Quantity
  • Availability
  • Add to Cart