ENA161208728 Enamine

N-(4-fluorophenyl)-2-[(propan-2-yl)amino]acetamide hydrochloride

  • MDL number MFCD07287174
  • Cas Number : 1049768-69-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09192

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09192
Availability details for
Quantity
  • Availability
  • Add to Cart