ENA161208750 Enamine

1-[(2-fluorophenyl)sulfonyl]piperazine hydrochloride

  • MDL number MFCD08445299
  • Cas Number : 1032757-97-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 270...272
Purity 95.00
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05031

Safety Information

msds https://www.enaminestore.com/catalog/EN300-05031
Availability details for
Quantity
  • Availability
  • Add to Cart