ENA161208760 Enamine

2-[(piperidin-1-yl)methyl]-3,4-dihydroquinazolin-4-one hydrochloride

  • MDL number MFCD08445331
  • Cas Number : 1049742-55-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09647

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09647
Availability details for
Quantity
  • Availability
  • Add to Cart