ENA161208763 Enamine

N-(2-ethylphenyl)-2-(methylamino)acetamide hydrochloride

  • MDL number MFCD07287723
  • Cas Number : 1049757-39-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 206...208
Purity 95.00
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09665

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09665
Availability details for
Quantity
  • Availability
  • Add to Cart