ENA161208766 Enamine

[3-(dimethylamino)propyl][(3-ethoxy-4-propoxyphenyl)methyl]amine dihydrochloride

  • MDL number MFCD07287724
  • Cas Number : 1049757-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09684

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09684
Availability details for
Quantity
  • Availability
  • Add to Cart