ENA161208776 Enamine

2-amino-N-(4-bromo-2-methylphenyl)acetamide hydrochloride

  • MDL number MFCD07287971
  • Cas Number : 1049767-05-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190...192
Purity 95.00
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10009

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10009
Availability details for
Quantity
  • Availability
  • Add to Cart