ENA161208789 Enamine

ethyl 2-(chloromethyl)-4-phenylquinoline-3-carboxylate hydrochloride

  • MDL number MFCD07308434
  • Cas Number : 1009306-55-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10126

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10126
Availability details for
Quantity
  • Availability
  • Add to Cart