ENA161208792 Enamine

{[3-methoxy-4-(prop-2-en-1-yloxy)phenyl]methyl}[(oxolan-2-yl)methyl]amine hydrochloride

  • MDL number MFCD07106386
  • Cas Number : 1048673-90-0
  • Molecular Weight : 313.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-10241

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10241
Availability details for
Quantity
  • Availability
  • Add to Cart