ENA161208802 Enamine

4-(chloromethyl)-N-(4-methoxyphenyl)-1,3-thiazol-2-amine hydrochloride

  • MDL number MFCD07288528
  • Cas Number : 1049746-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90-91
Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10432

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10432
Availability details for
Quantity
  • Availability
  • Add to Cart