ENA161208806 Enamine

2-{[(2-methoxyethyl)amino]methyl}quinazolin-4(3H)-one hydrochloride

  • MDL number MFCD07308562
  • Cas Number : 1049769-57-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158...160
Purity 95.00
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10443

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10443
Availability details for
Quantity
  • Availability
  • Add to Cart