ENA161208814 Enamine

(2-methoxyethyl)({[5-(thiophen-2-yl)-1,3,4-oxadiazol-2-yl]methyl})amine hydrochloride

  • MDL number MFCD06688754
  • Cas Number : 1052543-77-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145-152
Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-10550

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10550
Availability details for
Quantity
  • Availability
  • Add to Cart