ENA161208835 Enamine

4-[1-(3,4-dichlorophenyl)-2,5-dimethyl-1H-pyrrol-3-yl]-1,3-thiazol-2-amine hydrochloride

  • MDL number MFCD08447197
  • Cas Number : 1170210-77-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-10929

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10929
Availability details for
Quantity
  • Availability
  • Add to Cart