ENA161208839 Enamine

1-(2,6-dichlorobenzenesulfonyl)piperazine hydrochloride

  • MDL number MFCD07289054
  • Cas Number : 1049768-10-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11044

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11044
Availability details for
Quantity
  • Availability
  • Add to Cart