ENA161208846 Enamine

2-chloro-N-[2-(4-methoxyphenyl)-2-(piperidin-1-yl)ethyl]acetamide hydrochloride

  • MDL number MFCD07289417
  • Cas Number : 1049790-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11238

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11238
Availability details for
Quantity
  • Availability
  • Add to Cart