ENA161208854 Enamine

N'-(4-ethoxy-3-methoxybenzyl)-N,N-dimethylpropane-1,3-diamine dihydrochloride

  • MDL number MFCD07109534
  • Cas Number : 1049754-73-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 205...207
Purity 95.00
Salt Data 2HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11553

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11553
Availability details for
Quantity
  • Availability
  • Add to Cart