ENA161208855 Enamine

1-{3-[(diethylamino)methyl]-4-hydroxyphenyl}ethan-1-one hydrochloride

  • MDL number MFCD07289841
  • Cas Number : 1049743-37-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160-161
Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11627

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11627
Availability details for
Quantity
  • Availability
  • Add to Cart