ENA161208859 Enamine

N'-(2-chloroacetyl)-4-(diethylamino)benzohydrazide hydrochloride

  • MDL number MFCD07289945
  • Cas Number : 1052539-57-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-11664

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11664
Availability details for
Quantity
  • Availability
  • Add to Cart