ENA161208862 Enamine

1-{4-hydroxy-3-[(piperidin-1-yl)methyl]phenyl}ethan-1-one hydrochloride

  • MDL number MFCD07290462
  • Cas Number : 85843-13-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150-151
Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11722

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11722
Availability details for
Quantity
  • Availability
  • Add to Cart