ENA161208865 Enamine

benzyl(butan-2-yl)amine hydrochloride

  • MDL number MFCD07107483
  • Cas Number : 879652-85-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142-145
Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-11776

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11776
Availability details for
Quantity
  • Availability
  • Add to Cart