ENA161208870 Enamine

(2-phenylethyl)({[2-(prop-2-en-1-yloxy)phenyl]methyl})amine hydrochloride

  • MDL number MFCD07701113
  • Cas Number : 1052540-19-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11869

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11869
Availability details for
Quantity
  • Availability
  • Add to Cart