ENA161208873 Enamine

2-(1-adamantyl)ethanamine hydrochloride

  • MDL number MFCD00233047
  • Cas Number : 24644-08-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 329...331
Purity 95.00
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05618

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-05618
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart