ENA161208876 Enamine

4-(chloromethyl)-2-(4-ethylphenyl)-1,3-thiazole hydrochloride

  • MDL number MFCD07290507
  • Cas Number : 1049726-64-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 131...133
Purity 95.00
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12023

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12023
Availability details for
Quantity
  • Availability
  • Add to Cart