ENA161208877 Enamine

N-tert-butyl-2-(piperazin-1-yl)acetamide dihydrochloride

  • MDL number MFCD07290110
  • Cas Number : 1044707-29-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 35-36
Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12029

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12029
Availability details for
Quantity
  • Availability
  • Add to Cart