ENA161208878 Enamine

{[5-(4-chlorophenyl)furan-2-yl]methyl}(methyl)amine hydrochloride

  • MDL number MFCD07170081
  • Cas Number : 1049712-01-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12040

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12040
Availability details for
Quantity
  • Availability
  • Add to Cart