ENA161208882 Enamine

4-(chloromethyl)-2-(4-methoxyphenyl)-1,3-thiazole hydrochloride

  • MDL number MFCD07308421
  • Cas Number : 1052538-51-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83-84
Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12158

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12158
Availability details for
Quantity
  • Availability
  • Add to Cart