ENA161208883 Enamine

4-(chloromethyl)-2-[4-(propan-2-yl)phenyl]-1,3-thiazole hydrochloride

  • MDL number MFCD07287303
  • Cas Number : 1049790-37-5
  • Molecular Weight : 288.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-12159

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12159
Availability details for
Quantity
  • Availability
  • Add to Cart