ENA161208893 Enamine

3,4-dihydro-2H-1-benzothiopyran-4-amine hydrochloride

  • MDL number MFCD00035263
  • Cas Number : 15857-70-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 228-229
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-12532

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12532
Availability details for
Quantity
  • Availability
  • Add to Cart