ENA161208895 Enamine

5,6-dimethyl-2-[(piperazin-1-yl)methyl]-3H,4H-thieno[2,3-d]pyrimidin-4-one dihydrochloride

  • MDL number MFCD08447200
  • Cas Number : 1170919-73-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12538

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12538
Availability details for
Quantity
  • Availability
  • Add to Cart