ENA161208898 Enamine

2-[4-(2-chloroacetyl)piperazin-1-yl]-N-(propan-2-yl)acetamide hydrochloride

  • MDL number MFCD06673124
  • Cas Number : 1052531-90-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-12555

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12555
Availability details for
Quantity
  • Availability
  • Add to Cart