ENA161208916 Enamine

3-[(2-methoxyethyl)amino]-1lambda6-thiolane-1,1-dione hydrochloride

  • MDL number MFCD02340568
  • Cas Number : 483351-50-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-13006

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13006
Availability details for
Quantity
  • Availability
  • Add to Cart