ENA161208928 Enamine

3-(cyclopentylamino)-1lambda6-thiolane-1,1-dione hydrochloride

  • MDL number MFCD06685890
  • Cas Number : 1052545-34-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130-131
Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13183

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13183
Availability details for
Quantity
  • Availability
  • Add to Cart