ENA161208933 Enamine

[(3,4-dimethoxyphenyl)methyl](ethyl)amine hydrochloride

  • MDL number MFCD07106857
  • Cas Number : 1052544-93-0
  • Molecular Weight : 231.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139-140
Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-13217

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13217
Availability details for
Quantity
  • Availability
  • Add to Cart