ENA161208936 Enamine

N-[(2-butyl-1-benzofuran-3-yl)methyl]-N-methylamine hydrochloride

  • MDL number MFCD06671219
  • Cas Number : 1052552-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16...18
Purity 95.00
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13280

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13280
Availability details for
Quantity
  • Availability
  • Add to Cart